RIE240870084 Rieke Metals, Inc.

2-(4-Isopropoxybenzoyl)-4-methylpyridine

  • MDL number MFCD13152727
  • Molecular Weight : 255.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-isopropoxyphenyl)(4-methyl-2-pyridinyl)methanone
Inchi Code 1S/C16H17NO2/c1-11(2)19-14-6-4-13(5-7-14)16(18)15-10-12(3)8-9-17-15/h4-11H,1-3H3
Inchi Key QOLPYIZTPBLGEB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5614-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-33
Availability details for
Quantity
  • Availability
  • Add to Cart