RIE240870085 Rieke Metals, Inc.

2-(4-tert-Butylbenzoyl)-4-methylpyridine

  • MDL number MFCD13152728
  • Molecular Weight : 253.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5614-34

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-34
Availability details for
Quantity
  • Availability
  • Add to Cart