RIE240870092 Rieke Metals, Inc.

2-(4-Ethoxybenzoyl)-4-methylpyridine

  • MDL number MFCD13152735
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5614-36A

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-36A
Availability details for
Quantity
  • Availability
  • Add to Cart