RIE240870099 Rieke Metals, Inc.

2-(2,4-Difluorobenzoyl)-4-methylpyridine

  • MDL number MFCD13152742
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,4-difluorophenyl)(4-methyl-2-pyridinyl)methanone
Inchi Code 1S/C13H9F2NO/c1-8-4-5-16-12(6-8)13(17)10-3-2-9(14)7-11(10)15/h2-7H,1H3
Inchi Key IKVIEQWFBONHBP-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5614-38

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-38
Availability details for
Quantity
  • Availability
  • Add to Cart