RIE240870102 Rieke Metals, Inc.

2-(3,4-Difluorobenzoyl)-4-methylpyridine

  • MDL number MFCD13152745
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,4-difluorophenyl)(4-methyl-2-pyridinyl)methanone
Inchi Code 1S/C13H9F2NO/c1-8-4-5-16-12(6-8)13(17)9-2-3-10(14)11(15)7-9/h2-7H,1H3
Inchi Key QPGBVDPKDQKGOM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5614-41

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-41
Availability details for
Quantity
  • Availability
  • Add to Cart