RIE240870104 Rieke Metals, Inc.

2-(2,3-Dichlorobenzoyl)-4-methylpyridine

  • MDL number MFCD13152747
  • Molecular Weight : 266.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,3-dichlorophenyl)(4-methyl-2-pyridinyl)methanone
Inchi Code 1S/C13H9Cl2NO/c1-8-5-6-16-11(7-8)13(17)9-3-2-4-10(14)12(9)15/h2-7H,1H3
Inchi Key IAFCWCYCRLAABU-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5614-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-43
Availability details for
Quantity
  • Availability
  • Add to Cart