RIE240870105 Rieke Metals, Inc.

2-(2,4-Dichlorobenzoyl)-4-methylpyridine

  • MDL number MFCD13152748
  • Molecular Weight : 266.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5614-44

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-44
Availability details for
Quantity
  • Availability
  • Add to Cart