RIE240870106 Rieke Metals, Inc.

2-(2,5-Dichlorobenzoyl)-4-methylpyridine

  • MDL number MFCD13152749
  • Molecular Weight : 266.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,5-dichlorophenyl)(4-methyl-2-pyridinyl)methanone
Inchi Code 1S/C13H9Cl2NO/c1-8-4-5-16-12(6-8)13(17)10-7-9(14)2-3-11(10)15/h2-7H,1H3
Inchi Key IIKHHHUHJACOLM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5614-45

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-45
Availability details for
Quantity
  • Availability
  • Add to Cart