RIE240870113 Rieke Metals, Inc.

2-(2,6-Dimethoxybenzoyl)-4-methylpyridine

  • MDL number MFCD13152756
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5614-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-52
Availability details for
Quantity
  • Availability
  • Add to Cart