RIE240870115 Rieke Metals, Inc.

2-(3,5-Dimethoxybenzoyl)-4-methylpyridine

  • MDL number MFCD13152758
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5614-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-54
Availability details for
Quantity
  • Availability
  • Add to Cart