RIE240870117 Rieke Metals, Inc.

2-(2,4-Dimethylbenzoyl)-4-methylpyridine

  • MDL number MFCD13152760
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,4-dimethylphenyl)(4-methyl-2-pyridinyl)methanone
Inchi Code 1S/C15H15NO/c1-10-4-5-13(12(3)8-10)15(17)14-9-11(2)6-7-16-14/h4-9H,1-3H3
Inchi Key MUGYTRBJUZSZJB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5614-56

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5614-56
Availability details for
Quantity
  • Availability
  • Add to Cart