RIE240870124 Rieke Metals, Inc.

2-Benzoyl-3-methylpyridine

  • MDL number MFCD13152767
  • Molecular Weight : 197.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5615-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5615-01
Availability details for
Quantity
  • Availability
  • Add to Cart