RIE240870128 Rieke Metals, Inc.

2-(2-Methylbenzoyl)-3-methylpyridine

  • MDL number MFCD13152771
  • Molecular Weight : 211.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methylphenyl)(3-methyl-2-pyridinyl)methanone
Inchi Code 1S/C14H13NO/c1-10-6-3-4-8-12(10)14(16)13-11(2)7-5-9-15-13/h3-9H,1-2H3
Inchi Key SLDJUGFYTZKKGA-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5615-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5615-05
Availability details for
Quantity
  • Availability
  • Add to Cart