RIE240870134 Rieke Metals, Inc.

2-(2-Fluorobenzoyl)-3-methylpyridine

  • MDL number MFCD13152776
  • Molecular Weight : 215.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5615-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5615-11
Availability details for
Quantity
  • Availability
  • Add to Cart