RIE240870141 Rieke Metals, Inc.

2-(4-Bromobenzoyl)-3-methylpyridine

  • MDL number MFCD13152784
  • Molecular Weight : 276.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromophenyl)(3-methyl-2-pyridinyl)methanone
Inchi Code 1S/C13H10BrNO/c1-9-3-2-8-15-12(9)13(16)10-4-6-11(14)7-5-10/h2-8H,1H3
Inchi Key UKWNWDYPQAWINO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5615-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5615-19
Availability details for
Quantity
  • Availability
  • Add to Cart