RIE240870146 Rieke Metals, Inc.

2-(4-Cyanobenzoyl)-3-methylpyridine

  • MDL number MFCD13152789
  • Molecular Weight : 222.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5615-25

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5615-25
Availability details for
Quantity
  • Availability
  • Add to Cart