RIE240870154 Rieke Metals, Inc.

2-(4-Ethylbenzoyl)-3-methylpyridine

  • MDL number MFCD13152797
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-ethylphenyl)(3-methyl-2-pyridinyl)methanone
Inchi Code 1S/C15H15NO/c1-3-12-6-8-13(9-7-12)15(17)14-11(2)5-4-10-16-14/h4-10H,3H2,1-2H3
Inchi Key JSCRGIGYRFCVGM-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5615-35a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5615-35a
Availability details for
Quantity
  • Availability
  • Add to Cart