RIE240870155 Rieke Metals, Inc.

3-Methyl-2-(4-propylbenzoyl)pyridine

  • MDL number MFCD13152798
  • Molecular Weight : 239.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-methyl-2-pyridinyl)(4-propylphenyl)methanone
Inchi Code 1S/C16H17NO/c1-3-5-13-7-9-14(10-8-13)16(18)15-12(2)6-4-11-17-15/h4,6-11H,3,5H2,1-2H3
Inchi Key URYMUCIDHWZJRO-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5615-35b

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5615-35b
Availability details for
Quantity
  • Availability
  • Add to Cart