RIE240870159 Rieke Metals, Inc.

2-(4-Heptylbenzoyl)-3-methylpyridine

  • MDL number MFCD13152802
  • Molecular Weight : 295.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-heptylphenyl)(3-methyl-2-pyridinyl)methanone
Inchi Code 1S/C20H25NO/c1-3-4-5-6-7-10-17-11-13-18(14-12-17)20(22)19-16(2)9-8-15-21-19/h8-9,11-15H,3-7,10H2,1-2H3
Inchi Key KOSXLXVLKCMHRN-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5615-35f

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5615-35f
Availability details for
Quantity
  • Availability
  • Add to Cart