RIE240870164 Rieke Metals, Inc.

2-(4-Hexyloxybenzoyl)-3-methylpyridine

  • MDL number MFCD13152807
  • Molecular Weight : 297.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name [4-(hexyloxy)phenyl](3-methyl-2-pyridinyl)methanone
Inchi Code 1S/C19H23NO2/c1-3-4-5-6-14-22-17-11-9-16(10-12-17)19(21)18-15(2)8-7-13-20-18/h7-13H,3-6,14H2,1-2H3
Inchi Key DUMQJDZHXITPLT-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5615-36e

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5615-36e
Availability details for
Quantity
  • Availability
  • Add to Cart