RIE240870171 Rieke Metals, Inc.

2-(3,5-Difluorobenzoyl)-3-methylpyridine

  • MDL number MFCD13152814
  • Molecular Weight : 233.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-difluorophenyl)(3-methyl-2-pyridinyl)methanone
Inchi Code 1S/C13H9F2NO/c1-8-3-2-4-16-12(8)13(17)9-5-10(14)7-11(15)6-9/h2-7H,1H3
Inchi Key NFGJJDZUAOVUET-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5615-42

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5615-42
Availability details for
Quantity
  • Availability
  • Add to Cart