RIE240870172 Rieke Metals, Inc.

2-(2,3-Dichlorobenzoyl)-3-methylpyridine

  • MDL number MFCD13152815
  • Molecular Weight : 266.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,3-dichlorophenyl)(3-methyl-2-pyridinyl)methanone
Inchi Code 1S/C13H9Cl2NO/c1-8-4-3-7-16-12(8)13(17)9-5-2-6-10(14)11(9)15/h2-7H,1H3
Inchi Key KCGQSXMGFQPTAD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5615-43

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5615-43
Availability details for
Quantity
  • Availability
  • Add to Cart