RIE240870182 Rieke Metals, Inc.

2-(3,5-Dimethoxybenzoyl)-3-methylpyridine

  • MDL number MFCD13152826
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3,5-dimethoxyphenyl)(3-methyl-2-pyridinyl)methanone
Inchi Code 1S/C15H15NO3/c1-10-5-4-6-16-14(10)15(17)11-7-12(18-2)9-13(8-11)19-3/h4-9H,1-3H3
Inchi Key ATASHCHDUBFEOK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5615-54

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5615-54
Availability details for
Quantity
  • Availability
  • Add to Cart