RIE240870183 Rieke Metals, Inc.

2-(2,3-Dimethylbenzoyl)-3-methylpyridine

  • MDL number MFCD13152827
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,3-dimethylphenyl)(3-methyl-2-pyridinyl)methanone
Inchi Code 1S/C15H15NO/c1-10-6-4-8-13(12(10)3)15(17)14-11(2)7-5-9-16-14/h4-9H,1-3H3
Inchi Key HYARMSSTKNHLDY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5615-55

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5615-55
Availability details for
Quantity
  • Availability
  • Add to Cart