RIE240870186 Rieke Metals, Inc.

2-(2,6-Dimethylbenzoyl)-3-methylpyridine

  • MDL number MFCD13152830
  • Molecular Weight : 225.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,6-dimethylphenyl)(3-methyl-2-pyridinyl)methanone
Inchi Code 1S/C15H15NO/c1-10-6-4-7-11(2)13(10)15(17)14-12(3)8-5-9-16-14/h4-9H,1-3H3
Inchi Key WXUQUEIVIXWBQF-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5615-58

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5615-58
Availability details for
Quantity
  • Availability
  • Add to Cart