RIE240870189 Rieke Metals, Inc.

3-Methyl-2-(3,4,5-trifluorobenzoyl)pyridine

  • MDL number MFCD13152833
  • Molecular Weight : 251.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5615-61

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5615-61
Availability details for
Quantity
  • Availability
  • Add to Cart