RIE240870191 Rieke Metals, Inc.

2-Benzoyl-6-methoxypyridine

  • MDL number MFCD13152835
  • Molecular Weight : 213.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5617-01

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-01
Availability details for
Quantity
  • Availability
  • Add to Cart