RIE240870198 Rieke Metals, Inc.

6-Methoxy-2-(2-trifluoromethylbenzoyl)pyridine

  • MDL number MFCD13152842
  • Molecular Weight : 281.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5617-08

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-08
Availability details for
Quantity
  • Availability
  • Add to Cart