RIE240870199 Rieke Metals, Inc.

6-Methoxy-2-(3-trifluoromethylbenzoyl)pyridine

  • MDL number MFCD13152843
  • Molecular Weight : 281.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (6-methoxy-2-pyridinyl)[3-(trifluoromethyl)phenyl]methanone
Inchi Code 1S/C14H10F3NO2/c1-20-12-7-3-6-11(18-12)13(19)9-4-2-5-10(8-9)14(15,16)17/h2-8H,1H3
Inchi Key SKFBCBAHGPKTGY-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5617-09

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-09
Availability details for
Quantity
  • Availability
  • Add to Cart