RIE240870200 Rieke Metals, Inc.

6-Methoxy-2-(4-trifluoromethylbenzoyl)pyridine

  • MDL number MFCD13152844
  • Molecular Weight : 281.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5617-10

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-10
Availability details for
Quantity
  • Availability
  • Add to Cart