RIE240870201 Rieke Metals, Inc.

2-(2-Fluorobenzoyl)-6-methoxypyridine

  • MDL number MFCD13152845
  • Molecular Weight : 231.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-fluorophenyl)(6-methoxy-2-pyridinyl)methanone
Inchi Code 1S/C13H10FNO2/c1-17-12-8-4-7-11(15-12)13(16)9-5-2-3-6-10(9)14/h2-8H,1H3
Inchi Key MTEAYXGTJOOIAK-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5617-11

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-11
Availability details for
Quantity
  • Availability
  • Add to Cart