RIE240870205 Rieke Metals, Inc.

2-(3-Chlorobenzoyl)-6-methoxypyridine

  • MDL number MFCD13152849
  • Molecular Weight : 247.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chlorophenyl)(6-methoxy-2-pyridinyl)methanone
Inchi Code 1S/C13H10ClNO2/c1-17-12-7-3-6-11(15-12)13(16)9-4-2-5-10(14)8-9/h2-8H,1H3
Inchi Key KJTVXHXVASZGCD-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5617-15

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-15
Availability details for
Quantity
  • Availability
  • Add to Cart