RIE240870209 Rieke Metals, Inc.

2-(4-Bromobenzoyl)-6-methoxypyridine

  • MDL number MFCD13152853
  • Molecular Weight : 292.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5617-19

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-19
Availability details for
Quantity
  • Availability
  • Add to Cart