RIE240870213 Rieke Metals, Inc.

2-(3-Cyanobenzoyl)-6-methoxypyridine

  • MDL number MFCD13152857
  • Molecular Weight : 238.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5617-24

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-24
Availability details for
Quantity
  • Availability
  • Add to Cart