RIE240870216 Rieke Metals, Inc.

6-Methoxy-2-(4-phenoxybenzoyl)pyridine

  • MDL number MFCD13152860
  • Molecular Weight : 305.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5617-28

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-28
Availability details for
Quantity
  • Availability
  • Add to Cart