RIE240870220 Rieke Metals, Inc.

2-(4-Isopropoxybenzoyl)-6-methoxypyridine

  • MDL number MFCD13152864
  • Molecular Weight : 271.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-isopropoxyphenyl)(6-methoxy-2-pyridinyl)methanone
Inchi Code 1S/C16H17NO3/c1-11(2)20-13-9-7-12(8-10-13)16(18)14-5-4-6-15(17-14)19-3/h4-11H,1-3H3
Inchi Key HYSUYCYAZJYUJS-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5617-33

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-33
Availability details for
Quantity
  • Availability
  • Add to Cart