RIE240870224 Rieke Metals, Inc.

2-(4-Butylbenzoyl)-6-methoxypyridine

  • MDL number MFCD13152868
  • Molecular Weight : 269.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-butylphenyl)(6-methoxy-2-pyridinyl)methanone
Inchi Code 1S/C17H19NO2/c1-3-4-6-13-9-11-14(12-10-13)17(19)15-7-5-8-16(18-15)20-2/h5,7-12H,3-4,6H2,1-2H3
Inchi Key YVMNMUQDDFLAQV-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5617-35c

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-35c
Availability details for
Quantity
  • Availability
  • Add to Cart