RIE240870226 Rieke Metals, Inc.

2-(4-Hexylbenzoyl)-6-methoxypyridine

  • MDL number MFCD13152870
  • Molecular Weight : 297.40

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5617-35E

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-35E
Availability details for
Quantity
  • Availability
  • Add to Cart