RIE240870227 Rieke Metals, Inc.

2-(4-Heptylbenzoyl)-6-methoxypyridine

  • MDL number MFCD13152871
  • Molecular Weight : 311.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5617-35F

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-35F
Availability details for
Quantity
  • Availability
  • Add to Cart