RIE240870228 Rieke Metals, Inc.

2-(4-Ethoxybenzoyl)-6-methoxypyridine

  • MDL number MFCD13152872
  • Molecular Weight : 257.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-ethoxyphenyl)(6-methoxy-2-pyridinyl)methanone
Inchi Code 1S/C15H15NO3/c1-3-19-12-9-7-11(8-10-12)15(17)13-5-4-6-14(16-13)18-2/h4-10H,3H2,1-2H3
Inchi Key BVSYFHFCCYMJRB-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5617-36a

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-36a
Availability details for
Quantity
  • Availability
  • Add to Cart