RIE240870234 Rieke Metals, Inc.

2-(2,3-Difluorobenzoyl)-6-methoxypyridine

  • MDL number MFCD13152878
  • Molecular Weight : 249.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,3-difluorophenyl)(6-methoxy-2-pyridinyl)methanone
Inchi Code 1S/C13H9F2NO2/c1-18-11-7-3-6-10(16-11)13(17)8-4-2-5-9(14)12(8)15/h2-7H,1H3
Inchi Key OFWIHLDRFFWROX-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5617-37

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-37
Availability details for
Quantity
  • Availability
  • Add to Cart