RIE240870235 Rieke Metals, Inc.

2-(2,4-Difluorobenzoyl)-6-methoxypyridine

  • MDL number MFCD13152879
  • Molecular Weight : 249.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5617-38

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-38
Availability details for
Quantity
  • Availability
  • Add to Cart