RIE240870237 Rieke Metals, Inc.

2-(2,6-Difluorobenzoyl)-6-methoxypyridine

  • MDL number MFCD13152881
  • Molecular Weight : 249.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5617-40

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-40
Availability details for
Quantity
  • Availability
  • Add to Cart