RIE240870249 Rieke Metals, Inc.

2-(2,6-Dimethoxybenzoyl)-6-methoxypyridine

  • MDL number MFCD13152893
  • Molecular Weight : 273.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5617-52

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-52
Availability details for
Quantity
  • Availability
  • Add to Cart