RIE240870252 Rieke Metals, Inc.

2-(2,3-Dimethylbenzoyl)-6-methoxypyridine

  • MDL number MFCD13152896
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2,3-dimethylphenyl)(6-methoxy-2-pyridinyl)methanone
Inchi Code 1S/C15H15NO2/c1-10-6-4-7-12(11(10)2)15(17)13-8-5-9-14(16-13)18-3/h4-9H,1-3H3
Inchi Key CILPASKYFOLKQQ-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5617-55

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-55
Availability details for
Quantity
  • Availability
  • Add to Cart