RIE240870257 Rieke Metals, Inc.

2-(3,5-Dimethylbenzoyl)-6-methoxypyridine

  • MDL number MFCD13152901
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5617-60

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5617-60
Availability details for
Quantity
  • Availability
  • Add to Cart