RIE240870358 Rieke Metals, Inc.

4-(2-Methoxybenzoyl)isoquinoline

  • MDL number MFCD13153009
  • Molecular Weight : 263.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5620-02

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5620-02
Availability details for
Quantity
  • Availability
  • Add to Cart