RIE240870359 Rieke Metals, Inc.

4-(3-Methoxybenzoyl)isoquinoline

  • MDL number MFCD13153010
  • Molecular Weight : 263.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5620-03

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5620-03
Availability details for
Quantity
  • Availability
  • Add to Cart