RIE240870361 Rieke Metals, Inc.

4-(2-Methylbenzoyl)isoquinoline

  • MDL number MFCD13153012
  • Molecular Weight : 247.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://riekemetals.com/products-services/catnum/5620-05

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5620-05
Availability details for
Quantity
  • Availability
  • Add to Cart