RIE240870363 Rieke Metals, Inc.

4-(4-Methylbenzoyl)isoquinoline

  • MDL number MFCD13153014
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-isoquinolinyl(4-methylphenyl)methanone
Inchi Code 1S/C17H13NO/c1-12-6-8-13(9-7-12)17(19)16-11-18-10-14-4-2-3-5-15(14)16/h2-11H,1H3
Inchi Key KHKBAUCLCAPHQH-UHFFFAOYSA-N

Description

Purity 97%
Country of Origin US
Product Link http://riekemetals.com/products-services/catnum/5620-07

Safety Information

MSDS http://riekemetals.com/products-services/catnum/5620-07
Availability details for
Quantity
  • Availability
  • Add to Cart